Perfluoro(2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoic) acid 97%

Lieferant: Apollo Scientific
Achtung

PC450224-1G PC450224-5G
APOSPC450224-1GEA 260 EUR
APOSPC450224-1G APOSPC450224-5G
Perfluoro(2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoic) acid 97%
Perfluoro(2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoic) acid
Formel: C₁₈HF₃₅O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD03412273
CAS-Nummer: 52481-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR