5-Chloro-1,1-difluoro-3-oxapentane
Lieferant: Apollo Scientific
|
Achtung
|
PC450228-1G
PC450228-5G
APOSPC450228-1GEA
405
EUR
APOSPC450228-1G
APOSPC450228-5G
5-Chloro-1,1-difluoro-3-oxapentane
5-Chloro-1,1-difluoro-3-oxapentane
Formel:
C₄H₇ClF₂O Molecular Weight: 144,548 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD19442046 CAS-Nummer: 1309602-55-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...