3-Carbethoxy-2,6-bis(difluoromethyl)-4H-pyran-4-one
Lieferant: Apollo Scientific
PC450230-1G
PC450230-5G
APOSPC450230-1GEA
141
EUR
APOSPC450230-1G
APOSPC450230-5G
3-Carbethoxy-2,6-bis(difluoromethyl)-4H-pyran-4-one
3-Carbethoxy-2,6-bis(difluoromethyl)-4H-pyran-4-one
Formel:
C₁₀H₈F₄O₄ Molecular Weight: 268.162 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18074364 CAS-Nummer: 1309602-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...