2-[(2,3,4,5,6-Pentafluorophenyl)methoxy]phthalimide

Lieferant: Apollo Scientific
Achtung

PC450240-25G PC450240-5G
APOSPC450240-25GEA 458 EUR
APOSPC450240-25G APOSPC450240-5G
2-[(2,3,4,5,6-Pentafluorophenyl)methoxy]phthalimide
2-[(2,3,4,5,6-Pentafluorophenyl)methoxy]phthalimide
Formel: C₁₅H₆F₅NO₃
Molecular Weight: 343,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054391
CAS-Nummer: 57981-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR