3-Chloro-2,2,3,3-tetrafluoropropan-1-ol

Lieferant: Apollo Scientific
Achtung

PC450247-25G PC450247-1G PC450247-5G
APOSPC450247-25GEA 556 EUR
APOSPC450247-25G APOSPC450247-1G APOSPC450247-5G
3-Chloro-2,2,3,3-tetrafluoropropan-1-ol
3-Chloro-2,2,3,3-tetrafluoropropan-1-ol
Formel: C₃H₃ClF₄O
Molecular Weight: 166,502 g/mol
Siedepunkt: 107…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16621305
CAS-Nummer: 20411-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR