Ethyl (2R)-2-fluoropropanoate
Lieferant: Apollo Scientific
|
Achtung
|
PC450269-1G
PC450269-5G
APOSPC450269-1GEA
71.2
EUR
APOSPC450269-1G
APOSPC450269-5G
Ethyl (2R)-2-fluoropropanoate
Ethyl (2R)-2-fluoropropanoate
Formel:
C₅H₉FO₂ Molecular Weight: 120,122 g/mol Siedepunkt: 93…94 °C (300 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054290 CAS-Nummer: 72959-94-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...