Hexafluoroisopropyl methyl carbonate

Lieferant: Apollo Scientific

PC450407-25G PC450407-1G PC450407-5G
APOSPC450407-25GEA 1390 EUR
APOSPC450407-25G APOSPC450407-1G APOSPC450407-5G
Hexafluoroisopropyl methyl carbonate
Hexafluoroisopropyl methyl carbonate
Formel: C₅H₄F₆O₃
Molecular Weight: 226,074 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334384
CAS-Nummer: 607382-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR