N-Benzyl-3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanamide 95%

Lieferant: Apollo Scientific

PC450487-1G PC450487-250MG
APOSPC450487-1GEA 919 EUR
APOSPC450487-1G APOSPC450487-250MG
N-Benzyl-3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanamide 95%
N-Benzyl-3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanamide
Formel: C₁₁H₉F₆NO₂
Molecular Weight: 301.185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28901334
CAS-Nummer: 1980087-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR