2,2-Difluoroethyl chloroformate
Lieferant: Apollo Scientific
|
Gefahr
|
PC450490-1G
PC450490-5G
PC450490-25G
APOSPC450490-1GEA
126
EUR
APOSPC450490-1G
APOSPC450490-5G
APOSPC450490-25G
2,2-Difluoroethyl chloroformate
2,2-Difluoroethyl chloroformate
Formel:
C₃H₃ClF₂O₂ Molecular Weight: 144,505 g/mol Siedepunkt: 72…74 °C (150 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD24141311 CAS-Nummer: 111022-16-3 UN: 2742 ADR: 6.1+3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...