1,1,1-Trifluoro-2,2-bis(triethylsiloxy)ethane
Lieferant: Apollo Scientific
PC450516-1G
PC450516-5G
APOSPC450516-1GEA
248
EUR
APOSPC450516-1G
APOSPC450516-5G
1,1,1-Trifluoro-2,2-bis(triethylsiloxy)ethane
1,1,1-Trifluoro-2,2-bis(triethylsiloxy)ethane
Formel:
C₁₄H₃₁F₃O₂Si₂ Molecular Weight: 344,561 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30730303 CAS-Nummer: 1922958-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...