1,1,1-Trifluoro-3,3-bis(triethylsiloxy)propane

Lieferant: Apollo Scientific

PC450517-1G PC450517-5G
APOSPC450517-1GEA 219 EUR
APOSPC450517-1G APOSPC450517-5G
1,1,1-Trifluoro-3,3-bis(triethylsiloxy)propane
1,1,1-Trifluoro-3,3-bis(triethylsiloxy)propane
Formel: C₁₅H₃₃F₃O₂Si₂
Molecular Weight: 358.588 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30730304
CAS-Nummer: 2149598-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR