cis-4,4,4-Trifluoro-2-buten-1-ol

Lieferant: Apollo Scientific
Achtung

PC450521-1G PC450521-250MG PC450521-500MG
APOSPC450521-1GEA 763 EUR
APOSPC450521-1G APOSPC450521-250MG APOSPC450521-500MG
cis-4,4,4-Trifluoro-2-buten-1-ol
cis-4,4,4-Trifluoro-2-buten-1-ol
Formel: C₄H₅F₃O
Molecular Weight: 126,077 g/mol
Siedepunkt: 118,5 °C (60 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03092907
CAS-Nummer: 83706-98-3
UN: 1987
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR