3,3-Difluoro-1-nitro-2-propanol 95%

Lieferant: Apollo Scientific
Achtung

PC450554-1G PC450554-250MG
APOSPC450554-1GEA 1190 EUR
APOSPC450554-1G APOSPC450554-250MG
3,3-Difluoro-1-nitro-2-propanol 95%
3,3-Difluoro-1-nitro-2-propanol
Formel: C₃H₅F₂NO₃
Molecular Weight: 141,074 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28137318
CAS-Nummer: 148628-50-6
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR