Hexakis(3,3,3-trifluoropropoxy)phosphazene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC450559-1G
PC450559-250MG
APOSPC450559-1GEA
575
EUR
APOSPC450559-1G
APOSPC450559-250MG
Hexakis(3,3,3-trifluoropropoxy)phosphazene 95%
Hexakis(3,3,3-trifluoropropoxy)phosphazene
Formel:
C₁₈H₂₄F₁₈N₃O₆P₃ Molecular Weight: 813,292 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075243 CAS-Nummer: 1980062-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...