4-Iodo-1,1,2,2,2-pentafluoropentane

Lieferant: Apollo Scientific
Achtung

PC450569-25G PC450569-5G
APOSPC450569-25GEA 870 EUR
APOSPC450569-25G APOSPC450569-5G
4-Iodo-1,1,2,2,2-pentafluoropentane
4-Iodo-1,1,2,2,2-pentafluoropentane
Formel: C₅H₆F₅I
Molecular Weight: 287,998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18781884
CAS-Nummer: 1379295-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR