4,4'-Bis(trifluoromethyl)benzhydrol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Bis(4-(trifluoromethyl)phenyl)methanol
PC450579-1G
PC450579-250MG
APOSPC450579-1GEA
128
EUR
APOSPC450579-1G
APOSPC450579-250MG
4,4'-Bis(trifluoromethyl)benzhydrol
4,4'-Bis(trifluoromethyl)benzhydrol
Formel:
C₁₅H₁₀F₆O Molecular Weight: 320,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00626459 CAS-Nummer: 22543-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...