1,1,1-Trifluoro-3-iodopentane 97%

Lieferant: Apollo Scientific
Achtung

PC450584-1G PC450584-250MG
APOSPC450584-1GEA 323 EUR
APOSPC450584-1G APOSPC450584-250MG
1,1,1-Trifluoro-3-iodopentane 97%
1,1,1-Trifluoro-3-iodopentane
Formel: C₅H₈F₃I
Molecular Weight: 252,017 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28136066
CAS-Nummer: 75121-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR