2-Chloro-3,5-difluorobenzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(Bromomethyl)-2-chloro-3,5-difluorobenzene

PC45176-1G PC45176-250MG PC45176-5G
APOSPC45176-1GEA 575 EUR
APOSPC45176-1G APOSPC45176-250MG APOSPC45176-5G
2-Chloro-3,5-difluorobenzyl bromide 95%
2-Chloro-3,5-difluorobenzyl bromide
Formel: C₇H₄BrClF₂
Molecular Weight: 241,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28750503
CAS-Nummer: 1239353-49-1
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR