2-[1-Amino-2-(4-fluorophenoxy)ethylidene]malononitrile 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC4613-1G
PC4613-100MG
PC4613-5G
APOSPC4613-1GEA
285
EUR
APOSPC4613-1G
APOSPC4613-100MG
APOSPC4613-5G
2-[1-Amino-2-(4-fluorophenoxy)ethylidene]malononitrile 95%
2-[1-Amino-2-(4-fluorophenoxy)ethylidene]malononitrile
Formel:
C₁₁H₈FN₃O Molecular Weight: 217,199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00139200 CAS-Nummer: 1284228-23-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...