1-(2-Chloroethyl)-3-(2,4-difluorophenyl)urea
Lieferant: Apollo Scientific
|
Achtung
|
PC46361-1G
PC46361-250MG
PC46361-500MG
APOSPC46361-1GEA
323
EUR
APOSPC46361-1G
APOSPC46361-250MG
APOSPC46361-500MG
1-(2-Chloroethyl)-3-(2,4-difluorophenyl)urea
1-(2-Chloroethyl)-3-(2,4-difluorophenyl)urea
Formel:
C₉H₉ClF₂N₂O Molecular Weight: 234,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14586695 CAS-Nummer: 1248767-42-1 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...