2-Bromo-4-(trifluoromethyl)benzyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(chloromethyl)-4-(trifluoromethyl)benzene, 3-Bromo-4-(chloromethyl)benzotrifluoride

PC4733-1G PC4733-25G PC4733-5G
APOSPC4733-1GEA 23.2 EUR
APOSPC4733-1G APOSPC4733-25G APOSPC4733-5G
2-Bromo-4-(trifluoromethyl)benzyl chloride
2-Bromo-4-(trifluoromethyl)benzyl chloride
Formel: C₈H₅BrClF₃
Molecular Weight: 273.478 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03094249
CAS-Nummer: 480438-96-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR