1H,2H,2H-Perfluorooctanal hydrate (diol) 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctane-1,1-diol
PC47470-1G
PC47470-250MG
APOSPC47470-1GEA
667
EUR
APOSPC47470-1G
APOSPC47470-250MG
1H,2H,2H-Perfluorooctanal hydrate (diol) 97%
1H,2H,2H-Perfluorooctanal hydrate (diol)
Formel:
C₈H₅F₁₃O₂ Molecular Weight: 380,103 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31694621 CAS-Nummer: 2244084-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...