2,2,3,3-Tetrafluoro-1,4-benzodioxane-6-carbonitrile

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3,4-(Tetrafluoroethylenedioxy)benzonitrile, 2,2,3,3-Tetrafluoro-2,3-dihydro-1,4-benzodioxine-6-carbonitrile

PC4758-1G PC4758-250MG
APOSPC4758-1GEA 557 EUR
APOSPC4758-1G APOSPC4758-250MG
2,2,3,3-Tetrafluoro-1,4-benzodioxane-6-carbonitrile
2,2,3,3-Tetrafluoro-1,4-benzodioxane-6-carbonitrile
Formel: C₉H₃F₄NO₂
Molecular Weight: 233,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236310
CAS-Nummer: 215732-94-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR