5-Bromo-3,4-diaminophenylsulphur pentafluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-5-(pentafluorothio)benzene-1,2-diamine, 3-Bromo-5-(pentafluorosulphanyl)benzene-1,2-diamine

PC4807-1G PC4807-250MG
APOSPC4807-1GEA 348 EUR
APOSPC4807-1G APOSPC4807-250MG
5-Bromo-3,4-diaminophenylsulphur pentafluoride
5-Bromo-3,4-diaminophenylsulphur pentafluoride
Formel: C₆H₆BrF₅N₂S
Molecular Weight: 313,086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501036
CAS-Nummer: 1159512-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR