4,5,5,6,6,6-Hexafluoro-4-(trifluoromethyl)hexanoic acid 97%

Lieferant: Apollo Scientific
Gefahr

PC4807F-1G PC4807F-5G
APOSPC4807F-1GEA 33.5 EUR
APOSPC4807F-1G APOSPC4807F-5G
4,5,5,6,6,6-Hexafluoro-4-(trifluoromethyl)hexanoic acid 97%
4,5,5,6,6,6-Hexafluoro-4-(trifluoromethyl)hexanoic acid
Formel: C₇H₅F₉O₂
Molecular Weight: 292,099 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00155858
CAS-Nummer: 239463-95-7
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR