5-(Trifluoromethyl)-2-furaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Trifluoromethyl)furan-2-carboxaldehyde, 5-(Trifluoromethyl)furfural, 2-Formyl-5-(trifluoromethyl)furan
PC4825-1G
PC4825-250MG
APOSPC4825-1GEA
502
EUR
APOSPC4825-1G
APOSPC4825-250MG
5-(Trifluoromethyl)-2-furaldehyde 97%
5-(Trifluoromethyl)-2-furaldehyde
Formel:
C₆H₃F₃O₂ Molecular Weight: 164,082 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00525585 CAS-Nummer: 17515-80-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...