2-(4-Chloro-3-fluorophenoxy)acetic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC48289-1G
PC48289-250MG
PC48289-5G
APOSPC48289-1GEA
127
EUR
APOSPC48289-1G
APOSPC48289-250MG
APOSPC48289-5G
2-(4-Chloro-3-fluorophenoxy)acetic acid 95%
2-(4-Chloro-3-fluorophenoxy)acetic acid
Formel:
C₈H₆ClFO₃ Molecular Weight: 204.583 g/mol Schmelzpunkt: 132…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06823830 CAS-Nummer: 331-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...