2-[(Pentafluoropropanoyl)oxy]ethyl methacrylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[(2,2,3,3,3-Pentafluoropropanoyl)oxy]ethyl 2-methylprop-2-enoate, 2-(2,2,3,3,3-Pentafluoro-1-oxopropoxy)ethyl methacrylate

PC48330-25G PC48330-5G
APOSPC48330-25GEA 1210 EUR
APOSPC48330-25G APOSPC48330-5G
2-[(Pentafluoropropanoyl)oxy]ethyl methacrylate 97%
2-[(Pentafluoropropanoyl)oxy]ethyl methacrylate
Formel: C₉H₉F₅O₄
Molecular Weight: 276.157 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26407435
CAS-Nummer: 938459-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR