2-Bromo-3-fluorobenzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromo-3-fluorophenyl)methanol

PC48332-1G PC48332-5G PC48332-250MG
APOSPC48332-1GEA 89.9 EUR
APOSPC48332-1G APOSPC48332-5G APOSPC48332-250MG
2-Bromo-3-fluorobenzyl alcohol 97%
2-Bromo-3-fluorobenzyl alcohol
Formel: C₇H₆BrFO
Molecular Weight: 205,024 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12911577
CAS-Nummer: 1184915-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR