2-Fluoro-5-methoxyphenylacetic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Carboxymethyl)-4-fluoroanisole, (2-Fluoro-5-methoxyphenyl)acetic acid, 2-(2-Fluoro-5-methoxyphenyl)acetic acid

PC48341-1G PC48341-250MG PC48341-5G PC48341-25G
APOSPC48341-1GEA 46.4 EUR
APOSPC48341-1G APOSPC48341-250MG APOSPC48341-5G APOSPC48341-25G
2-Fluoro-5-methoxyphenylacetic acid 97%
2-Fluoro-5-methoxyphenylacetic acid
Formel: C₉H₉FO₃
Molecular Weight: 184,164 g/mol
Schmelzpunkt: 81…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13185602
CAS-Nummer: 798563-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR