7-Fluoro-1,3-benzoxazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Fluorobenzo[d]oxazole
PC48343-1G
PC48343-250MG
PC48343-5G
APOSPC48343-1GEA
464
EUR
APOSPC48343-1G
APOSPC48343-250MG
APOSPC48343-5G
7-Fluoro-1,3-benzoxazole 95%
7-Fluoro-1,3-benzoxazole
Formel:
C₇H₄FNO Schmelzpunkt: 43…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD19374434 CAS-Nummer: 1267061-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...