2-Bromo-5-fluorobenzo[b]furan-3(2H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-fluorocoumaran-3-one, 2-Bromo-5-fluoro-1-benzofuran-3(2H)-one, 2-Bromo-2,3-dihydro-5-fluoro-3-oxo-1-benzofuran

PC48347-1G PC48347-250MG
APOSPC48347-1GEA 713 EUR
APOSPC48347-1G APOSPC48347-250MG
2-Bromo-5-fluorobenzo[b]furan-3(2H)-one
2-Bromo-5-fluorobenzo[b]furan-3(2H)-one
Formel: C₈H₄BrFO₂
Molecular Weight: 231.019 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28096612
CAS-Nummer: 1823950-80-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR