3,4-dichloro-3,4,4-trifluorobut-1-ene

Lieferant: Apollo Scientific
Achtung

PC4835-25G PC4835-5G
APOSPC4835-25GEA 919 EUR
APOSPC4835-25G APOSPC4835-5G
3,4-dichloro-3,4,4-trifluorobut-1-ene
3,4-dichloro-3,4,4-trifluorobut-1-ene
Formel: C₄H₃Cl₂F₃
Molecular Weight: 178.968 g/mol
Siedepunkt: 79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00155750
CAS-Nummer: 374-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR