2'-Hydroxy-2,2,2,6'-tetrafluoroacetophenone 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluoro-6-hydroxyphenyl)-2,2,2-trifluoroethan-1-one, 3-Fluoro-2-(trifluoroacetyl)phenol

PC48355-1G PC48355-250MG PC48355-5G
APOSPC48355-1GEA 232 EUR
APOSPC48355-1G APOSPC48355-250MG APOSPC48355-5G
2'-Hydroxy-2,2,2,6'-tetrafluoroacetophenone 95%
2'-Hydroxy-2,2,2,6'-tetrafluoroacetophenone
Formel: C₈H₄F₄O₂
Molecular Weight: 208.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25541879
CAS-Nummer: 1445995-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR