7-Bromo-6-fluoro-1,3-benzoxazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Bromo-6-fluorobenzo[d]oxazole
PC48357-1G
PC48357-250MG
APOSPC48357-1GEA
551
EUR
APOSPC48357-1G
APOSPC48357-250MG
7-Bromo-6-fluoro-1,3-benzoxazole 97%
7-Bromo-6-fluoro-1,3-benzoxazole
Formel:
C₇H₃BrFNO Molecular Weight: 216.007 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25541880 CAS-Nummer: 1445995-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...