2-(Difluoromethyl)benzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(Bromomethyl)benzal fluoride, 1-(Bromomethyl)-2-(difluoromethyl)benzene
PC48365-1G
PC48365-250MG
APOSPC48365-1GEA
549
EUR
APOSPC48365-1G
APOSPC48365-250MG
2-(Difluoromethyl)benzyl bromide
2-(Difluoromethyl)benzyl bromide
Formel:
C₈H₇BrF₂ Molecular Weight: 221.042 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18791598 CAS-Nummer: 1222556-61-7 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...