3-Bromo-2-chloro-6-fluorobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Bromo-2-chloro-6-fluorophenyl)methanol

PC48378-1G PC48378-5G
APOSPC48378-1GEA 58 EUR
APOSPC48378-1G APOSPC48378-5G
3-Bromo-2-chloro-6-fluorobenzyl alcohol
3-Bromo-2-chloro-6-fluorobenzyl alcohol
Formel: C₇H₅BrClFO
Molecular Weight: 239,469 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20923583
CAS-Nummer: 1784628-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR