2'-Fluoro-3'-methylacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluoro-3-methylphenyl)ethanone

PC48390-1G PC48390-5G PC48390-25G
APOSPC48390-1GEA 52.2 EUR
APOSPC48390-1G APOSPC48390-5G APOSPC48390-25G
2'-Fluoro-3'-methylacetophenone
2'-Fluoro-3'-methylacetophenone
Formel: C₉H₉FO
Molecular Weight: 152.166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194343
CAS-Nummer: 865664-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR