3,4,5-Trifluorostyrene 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2,3-Trifluoro-5-vinylbenzene, 5-Ethenyl-1,2,3-trifluorobenzene

PC48409-1G PC48409-25G PC48409-5G
APOSPC48409-1GEA 79.9 EUR
APOSPC48409-1G APOSPC48409-25G APOSPC48409-5G
3,4,5-Trifluorostyrene 95%
3,4,5-Trifluorostyrene
Formel: C₈H₅F₃
Molecular Weight: 158.121 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18632531
CAS-Nummer: 159294-99-2
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR