5-(Hydroxymethyl)-2-(trifluoromethyl)-1,3-thiazole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-(Trifluoromethyl)thiazol-5-yl)methanol, [2-(Trifluoromethyl)-1,3-thiazol-5-yl]methanol
PC48418-250MG
PC48418-1G
PC48418-5G
APOSPC48418-250MGEA
166
EUR
APOSPC48418-250MG
APOSPC48418-1G
APOSPC48418-5G
5-(Hydroxymethyl)-2-(trifluoromethyl)-1,3-thiazole 95+%
5-(Hydroxymethyl)-2-(trifluoromethyl)-1,3-thiazole
Formel:
C₅H₄F₃NOS Molecular Weight: 183,152 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18632535 CAS-Nummer: 131748-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...