2,4,6-Trifluorobenzyl methacrylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4,6-Trifluorobenzyl 2-methylprop-2-enoate
PC48444-1G
PC48444-5G
PC48444-25G
APOSPC48444-1GEA
134
EUR
APOSPC48444-1G
APOSPC48444-5G
APOSPC48444-25G
2,4,6-Trifluorobenzyl methacrylate
2,4,6-Trifluorobenzyl methacrylate
Formel:
C₁₁H₉F₃O₂ Molecular Weight: 230.183 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28096613 CAS-Nummer: 1820739-95-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...