6'-Bromo-3'-chloro-2'-fluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(6-Bromo-3-chloro-2-fluorophenyl)ethanone, 1-(6-Bromo-3-chloro-2-fluorophenyl)ethan-1-one

PC48451-1G PC48451-250MG
APOSPC48451-1GEA 154 EUR
APOSPC48451-1G APOSPC48451-250MG
6'-Bromo-3'-chloro-2'-fluoroacetophenone
6'-Bromo-3'-chloro-2'-fluoroacetophenone
Formel: C₈H₅BrClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD28096615
CAS-Nummer: 1540199-39-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR