6-Bromo-3-chloro-2-fluorobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Bromo-3-chloro-2-fluorophenyl)methanol

PC48465-1G PC48465-250MG PC48465-5G
APOSPC48465-1GEA 206 EUR
APOSPC48465-1G APOSPC48465-250MG APOSPC48465-5G
6-Bromo-3-chloro-2-fluorobenzyl alcohol
6-Bromo-3-chloro-2-fluorobenzyl alcohol
Formel: C₇H₅BrClFO
Molecular Weight: 239,469 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24848703
CAS-Nummer: 1449008-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR