2-Fluoroterephthalic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluorobenzene-1,4-dicarboxylic acid

PC48471-250MG PC48471-1G
APOSPC48471-250MGEA 27.7 EUR
APOSPC48471-250MG APOSPC48471-1G
2-Fluoroterephthalic acid 95+%
2-Fluoroterephthalic acid
Formel: C₈H₅FO₄
Molecular Weight: 184,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407274
CAS-Nummer: 3906-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR