(3,5-Dichloro-2-fluorophenyl)boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dichloro-2-fluorophenylboronic acid

PC48478-1G PC48478-500MG
APOSPC48478-1GEA 435 EUR
APOSPC48478-1G APOSPC48478-500MG
(3,5-Dichloro-2-fluorophenyl)boronic acid
(3,5-Dichloro-2-fluorophenyl)boronic acid
Formel: C₆H₄BCl₂FO₂
Molecular Weight: 208,81 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28786331
CAS-Nummer: 2048237-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR