4-(Pentafluoroethyl)benzonitrile 95%
Lieferant: Apollo Scientific
PC48485-1G
PC48485-500MG
PC48485-5G
APOSPC48485-1GEA
359
EUR
APOSPC48485-1G
APOSPC48485-500MG
APOSPC48485-5G
4-(Pentafluoroethyl)benzonitrile 95%
4-(Pentafluoroethyl)benzonitrile
Formel:
C₉H₄F₅N Molecular Weight: 221.127 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19439995 CAS-Nummer: 128273-61-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...