(1S)-1-Amino-4-fluoroindane hydrochloride 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1S)-2,3-Dihydro-4-fluoro-1H-inden-1-amine hydrochloride, (S)-4-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride
PC48531-100MG
PC48531-1G
PC48531-250MG
APOSPC48531-100MGEA
123
EUR
APOSPC48531-100MG
APOSPC48531-1G
APOSPC48531-250MG
(1S)-1-Amino-4-fluoroindane hydrochloride 95+%
(1S)-1-Amino-4-fluoroindane hydrochloride
Formel:
C₉H₁₀FN·ClH Molecular Weight: 187,642 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16295140 CAS-Nummer: 1286734-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...