6-Bromo-3-chloro-2-fluoroaniline, N-BOC protected
Lieferant: Apollo Scientific
Achtung
Synonyme: tert-Butyl (6-bromo-3-chloro-2-fluorophenyl)carbamate
PC48558-1G PC48558-250MG
APOSPC48558-1GEA 847 EUR
APOSPC48558-1G APOSPC48558-250MG
6-Bromo-3-chloro-2-fluoroaniline, N-BOC protected
6-Bromo-3-chloro-2-fluoroaniline, N-BOC protected
Formel: C₁₁H₁₂BrClFNO₂
Molecular Weight: 324,574 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28096623
CAS-Nummer: 1820705-04-1