3-Amino-2,2-difluoropropan-1-ol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2-Difluoro-3-hydroxypropan-1-amine
PC48564-1G
PC48564-250MG
PC48564-5G
APOSPC48564-1GEA
232
EUR
APOSPC48564-1G
APOSPC48564-250MG
APOSPC48564-5G
3-Amino-2,2-difluoropropan-1-ol 97%
3-Amino-2,2-difluoropropan-1-ol
Formel:
C₃H₇F₂NO Molecular Weight: 111,091 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19207959 CAS-Nummer: 155310-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...