Ethyl 2-bromo-3,3,3-trifluoropropanoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-bromo-3,3,3-trifluoropropionate

PC48576-1G PC48576-5G
APOSPC48576-1GEA 125 EUR
APOSPC48576-1G APOSPC48576-5G
Ethyl 2-bromo-3,3,3-trifluoropropanoate
Ethyl 2-bromo-3,3,3-trifluoropropanoate
Formel: C₅H₆BrF₃O₂
Molecular Weight: 234,999 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18641866
CAS-Nummer: 108221-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR